Learn More
United Scientific™ Set of 12 Electrodes, Aluminum, Copper, Tin, Iron, Zinc, and Carbon (2 each)
Convenient set of commonly used electrodes
Supplier:  United Scientific™ UNELCSET
Catalog No. S01304
Only null left
Explore available promotions
Alert:
Prop 65
Description
Description
Use these flat, rigid electrodes in a variety of electrochemical demonstrations and experiments. The 127 mm long strips can fit into a variety of beakers, wet cells, and porous cups. If needed, the top can be bent over to stand upright in a container. Since the strips consist of one type of metal, they can be used in electroplating demonstrations. Also available as a set of 12 electrodes (two each of Aluminum, Carbon, Copper, Iron, Tin, and Zinc).
  • Available in six different metals
  • 127 mm x 19 mm x 1.2 mm thick
Specifications
Specifications
Electric Wet Cell
Product Suggestions
Product Suggestions