Learn More
Sesame Oil, NF, Spectrum™ Chemical
SE130, 8008-74-0
$1566.80 - $10819.77
Chemical Identifiers
CAS 8008-74-0
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18606187
View Documents
Spectrum Chemical Mfg Cor
SE130500MLGLCS6
6 x 500 mL Amber Glass Bottle
Case of 6 Each for $1,566.80
Only null left
18606189
View Documents
Spectrum Chemical Mfg Cor
SE1304LTGLCS4
4 x 4 L Amber Glass Bottle
Case of 4 Each for $4,937.69
Only null left
18606191
View Documents
Spectrum Chemical Mfg Cor
SE130200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Spectrum™ Chemical Sesame Oil, NF is used as an excipient solvent carrier in suspensions and emulsions, ointments, liniments, ophthalmic preparations, oral capsules and suppositories. All Spectrum Chemical NF are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Chemical Identifiers
8008-74-0
Specifications
8008-74-0
Amber Glass Bottle
188-195
1
6 x 500 mL
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'