Learn More
- Home
- Products
- Support Services and Compliance Services
- Safety Subscription Program Services
- Lone Worker Monitoring Services
- Blackline Safety™ Service for G7 Connected Safety Monitor with Multi-Gas Sensor Cartridge, NH3, CO2
Blackline Safety™ Service for G7 Connected Safety Monitor with Multi-Gas Sensor Cartridge, NH3, CO2
Catalog No. p-7160619
Change view
Click to view available options
Service Offering:
G7 Insight
Self-monitored with voice
Self-monitored, no voice
SOC-monitored with voice
SOC-monitored, no voice
Duration:
1 Year
2 Years
3 Years
4 Years
5 Years
25 product options available for selection
| Catalog No. | Service Offering | Duration |
|---|---|---|
| 170001611 | G7 Insight | 1 Year |
| 170001684 | G7 Insight | 2 Years |
| 170001757 | G7 Insight | 3 Years |
| 170001830 | G7 Insight | 4 Years |
| 170001903 | - | 5 Years |
| 170001246 | Self-monitored, no voice | - |
| 170001319 | Self-monitored, no voice | 2 Years |
| 170001392 | Self-monitored, no voice | 3 Years |
| 170001465 | Self-monitored, no voice | 4 Years |
| 170001538 | Self-monitored, no voice | 5 Years |
| 170000881 | - | - |
| 170000954 | Self-monitored with voice | 2 Years |
| 170001027 | - | 3 Years |
| 170001100 | Self-monitored with voice | 4 Years |
| 170001173 | Self-monitored with voice | 5 Years |
| 170000516 | SOC-monitored, no voice | 1 Year |
| 170000589 | SOC-monitored, no voice | 2 Years |
| 170000662 | SOC-monitored, no voice | 3 Years |
| 170000735 | SOC-monitored, no voice | - |
| 170000808 | SOC-monitored, no voice | 5 Years |
| 170000151 | - | 1 Year |
| 170000224 | SOC-monitored with voice | 2 Years |
| 170000297 | SOC-monitored with voice | - |
| 170000370 | SOC-monitored with voice | 4 Years |
| 170000443 | SOC-monitored with voice | 5 Years |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
Description
Optional services leverage G7's connectivity to link employees with monitoring personnel.
- Real-time Alerts: Required for both self-monitored and Blackline-monitored customers. G7 devices can be configured to detect incidents and signal for help as they happen by notifying live monitoring personnel.
- Safety Operations Center (SOC): Blackline's inhouse SOC focuses on monitoring the wellbeing of employees.Personnel manage all alerts in real-time.
- Two-way Voice Calling: G7c users can instantly connect with monitoring personnel via G7c's built-in two-way speakerphone. Two-way text messaging is also available for both G7c and G7x users.
- Push-to-talk (PTT): New G7c devices feature PTT functionality. It allows the G7c to function similar to a walkie-talkie.
Specifications
Specifications
| For Use With (Equipment) | G7 Multi-gas Cartridge, NH3, CO2 |
| Duration | 1 Year |
| Product Type | Service |
| Service Offering | G7 Insight |