Learn More
Blackline Safety™ Service for G7 Connected Safety Monitor with Multi-Gas Sensor Cartridge, CO2, CO, O2
Catalog No. 170001629
Change view
Click to view available options
Service Offering:
G7 Insight
Self-monitored with voice
Self-monitored, no voice
SOC-monitored with voice
SOC-monitored, no voice
Duration:
1 Year
2 Years
3 Years
4 Years
5 Years
25 product options available for selection
Product selection table with 25 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Service Offering Duration
170001629 G7 Insight 1 Year
170001702 G7 Insight 2 Years
170001775 G7 Insight 3 Years
170001848 G7 Insight 4 Years
170001921 G7 Insight 5 Years
170001264 Self-monitored, no voice 1 Year
170001337 - 2 Years
170001410 Self-monitored, no voice 3 Years
170001483 Self-monitored, no voice 4 Years
170001556 Self-monitored, no voice 5 Years
170000899 Self-monitored with voice 1 Year
170000972 - 2 Years
170001045 Self-monitored with voice 3 Years
170001118 Self-monitored with voice -
170001191 Self-monitored with voice 5 Years
170000534 SOC-monitored, no voice 1 Year
170000607 SOC-monitored, no voice 2 Years
170000680 SOC-monitored, no voice 3 Years
170000753 SOC-monitored, no voice -
170000826 SOC-monitored, no voice 5 Years
170000169 SOC-monitored with voice 1 Year
170000242 SOC-monitored with voice 2 Years
170000315 SOC-monitored with voice 3 Years
170000388 SOC-monitored with voice 4 Years
170000461 SOC-monitored with voice -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
25 options
Catalog No. 170001629 Supplier Blackline Safety™ Supplier No. SERXXQCMOX1Y
Blackline Safety™'s G7 service plans can help you confidently identify the real-time whereabouts and wellbeing of workers in the field.
Optional services leverage G7's connectivity to link employees with monitoring personnel.
  • Real-time Alerts: Required for both self-monitored and Blackline-monitored customers. G7 devices can be configured to detect incidents and signal for help as they happen by notifying live monitoring personnel.
  • Safety Operations Center (SOC): Blackline's inhouse SOC focuses on monitoring the wellbeing of employees.Personnel manage all alerts in real-time.
  • Two-way Voice Calling: G7c users can instantly connect with monitoring personnel via G7c's built-in two-way speakerphone. Two-way text messaging is also available for both G7c and G7x users.
  • Push-to-talk (PTT): New G7c devices feature PTT functionality. It allows the G7c to function similar to a walkie-talkie.
Specifications
For Use With (Equipment) G7 Multi-gas Cartridge, CO2, CO, O2
Duration 1 Year
Product Type Service
Service Offering G7 Insight