Learn More
Blackline Safety™ Service for G7 Connected Safety Monitor with Multi-Gas Sensor Cartridge, CO2, CO/H2S Twin Tox, LEL-I, O2
Blackline Safety™'s G7 service plans can help you confidently identify the real-time whereabouts and wellbeing of workers in the field.
Supplier: Blackline Safety™ SERSXQCDIO2Y
Description
Optional services leverage G7's connectivity to link employees with monitoring personnel.
- Real-time Alerts: Required for both self-monitored and Blackline-monitored customers. G7 devices can be configured to detect incidents and signal for help as they happen by notifying live monitoring personnel.
- Safety Operations Center (SOC): Blackline's inhouse SOC focuses on monitoring the wellbeing of employees.Personnel manage all alerts in real-time.
- Two-way Voice Calling: G7c users can instantly connect with monitoring personnel via G7c's built-in two-way speakerphone. Two-way text messaging is also available for both G7c and G7x users.
- Push-to-talk (PTT): New G7c devices feature PTT functionality. It allows the G7c to function similar to a walkie-talkie.
Chemical Identifiers
| G7 Multi-gas Cartridge, CO2, CO/H2S Twin Tox, LEL-I, O2 |
Specifications
| G7 Multi-gas Cartridge, CO2, CO/H2S Twin Tox, LEL-I, O2 | |
| 2 Years | |
| Self-monitored, no voice |