Learn More
Blackline Safety™ Service for G7 Connected Safety Monitor with Single-Gas Sensor Cartridge, SO2
Catalog No. 170001673
Change view
Click to view available options
Service Offering:
G7 Insight
Self-monitored with voice
Self-monitored, no voice
SOC-monitored with voice
SOC-monitored, no voice
Duration:
1 Year
2 Years
3 Years
4 Years
5 Years
25 product options available for selection
Product selection table with 25 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Service Offering Duration
170001673 G7 Insight 1 Year
170001746 - 2 Years
170001819 G7 Insight 3 Years
170001892 G7 Insight 4 Years
170001965 G7 Insight 5 Years
170001308 Self-monitored, no voice 1 Year
170001381 Self-monitored, no voice 2 Years
170001454 - 3 Years
170001527 Self-monitored, no voice -
170001600 Self-monitored, no voice 5 Years
170000943 Self-monitored with voice 1 Year
170001016 Self-monitored with voice 2 Years
170001089 Self-monitored with voice 3 Years
170001162 Self-monitored with voice 4 Years
170001235 Self-monitored with voice 5 Years
170000578 - 1 Year
170000651 SOC-monitored, no voice 2 Years
170000724 SOC-monitored, no voice 3 Years
170000797 SOC-monitored, no voice 4 Years
170000870 SOC-monitored, no voice 5 Years
170000213 SOC-monitored with voice 1 Year
170000286 SOC-monitored with voice 2 Years
170000359 SOC-monitored with voice 3 Years
170000432 SOC-monitored with voice 4 Years
170000505 SOC-monitored with voice 5 Years
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
25 options
Catalog No. 170001673 Supplier Blackline Safety™ Supplier No. SERXXSS1Y
Blackline Safety™'s G7 service plans can help you confidently identify the real-time whereabouts and wellbeing of workers in the field.
Optional services leverage G7's connectivity to link employees with monitoring personnel.
  • Real-time Alerts: Required for both self-monitored and Blackline-monitored customers. G7 devices can be configured to detect incidents and signal for help as they happen by notifying live monitoring personnel.
  • Safety Operations Center (SOC): Blackline's inhouse SOC focuses on monitoring the wellbeing of employees.Personnel manage all alerts in real-time.
  • Two-way Voice Calling: G7c users can instantly connect with monitoring personnel via G7c's built-in two-way speakerphone. Two-way text messaging is also available for both G7c and G7x users.
  • Push-to-talk (PTT): New G7c devices feature PTT functionality. It allows the G7c to function similar to a walkie-talkie.
Specifications
For Use With (Equipment) G7 Single-gas Cartridge, SO2
Duration 1 Year
Service Offering G7 Insight