Learn More
METTLER TOLEDO™ Service for Excellence XPR Micro and Micro-Analytical Balances
Catalog No. 01670009
Change view
Click to view available options
Includes:
EQPac Regulated Balances with USP
IPac Balances with ACC
StarterPac Installation with ACC
For Use With (Equipment):
Micro/Micro XPR Analytical Balances
XPR2U Ultra Micro Balances
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes For Use With (Equipment)
01670009 StarterPac Installation with ACC XPR2U Ultra Micro Balances
01670011 EQPac Regulated Balances with USP Micro/Micro XPR Analytical Balances
01670021 IPac Balances with ACC Micro/Micro XPR Analytical Balances
01670034 StarterPac Installation with ACC Micro/Micro XPR Analytical Balances
01670039 EQPac Regulated Balances with USP XPR2U Ultra Micro Balances
01670045 IPac Balances with ACC XPR2U Ultra Micro Balances
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01670009 Supplier METTLER TOLEDO™ Supplier No. B99474110111
Professional installation completed by a trained service technician will guarantee maximum performance and optimal use.
To achieve peak performance and compliance with quality systems and regulatory requirements, take the right steps at the beginning of equipment use. The Accuracy Calibration Certificate (ACC) is the new benchmark in the calibration of weighing equipment. Tests include repeatability, eccentricity and error of indication. A statement of measurement uncertainty is also included.
  • StarterPac includes: Professional installation performed by a trained and authorized service technician, installation confirmation/documentation, immediate deployment and use of your equipment, professionally trained operating personnel
  • IPac includes: Documented standard qualification and initial calibration, compliance with internal and external regulations, immediate deployment for quick, safe release to routine operation, and improved productivity with correct equipment set-up and initial user training
  • EQPac includes: Regulatory conformance qualification documentation, professional qualification including test procedures, specification of weighing ranges and calibration, immediate deployment for quick, safe release to begin routine operation, improved productivity with correct equipment set-up and initial user training, cost savings through the use of a manufacturer developed qualification protocol
Specifications
For Use With (Equipment) XPR2U Ultra Micro Balances
Duration 1 Day
Product Type Balance Service
Product Line Excellence
Service Offering Installation and Customer Training
Includes StarterPac Installation with ACC