Learn More
METTLER TOLEDO™ Service for Excellence XPR Balances
Professional installation completed by a trained service technician will guarantee maximum performance and optimal use.
Supplier:  METTLER TOLEDO™ B99484110109
 View more versions of this product
Catalog No. 01670035
Only null left
Description
Description
To achieve peak performance and compliance with quality systems and regulatory requirements, take the right steps at the beginning of equipment use. The Accuracy Calibration Certificate (ACC) is the new benchmark in the calibration of weighing equipment. Tests include repeatability, eccentricity and error of indication. A statement of measurement uncertainty is also included.
  • StarterPac includes: Professional installation performed by a trained and authorized service technician, installation confirmation/documentation, immediate deployment and use of your equipment, professionally trained operating personnel
  • IPac includes: Documented standard qualification and initial calibration, compliance with internal and external regulations, immediate deployment for quick, safe release to routine operation, and improved productivity with correct equipment set-up and initial user training
  • EQPac includes: Regulatory conformance qualification documentation, professional qualification including test procedures, specification of weighing ranges and calibration, immediate deployment for quick, safe release to begin routine operation, improved productivity with correct equipment set-up and initial user training, cost savings through the use of a manufacturer developed qualification protocol
Specifications
Chemical Identifiers
1 Day
Balance Service
Specifications
Excellence XPR Analytical Balances
Balance Service
IPac Balances with Accredited ACC
1 Day
Installation Qualification
Product Suggestions
Product Suggestions