missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One VACUETTE™ CAT Serum Separator Clot Activator Blood Collection Tubes

Catalog No. 22040542
Change view
Click to view available options
Capacity (Metric):
2.5 mL
3.5 mL
5 mL
8 mL
Closure Color:
Gold with Gold Ring
Red with White Ring
Red with Yellow Ring
Tube Size:
13 x 75 mm
13 x 100 mm
16 x 100 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color Tube Size
22040542 2.5 mL Red with White Ring 13 x 75 mm
22040536 3.5 mL Red with Yellow Ring 13 x 75 mm
22040541 5 mL Red with Yellow Ring 13 x 100 mm
22040546 3.5 mL Gold with Gold Ring 13 x 75 mm
22040537 8 mL Red with Yellow Ring 16 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22040542 Supplier Greiner Bio-One Supplier No. 454243P

missing translation for 'validMainframeMsg'

VACUETTE™ CAT Serum Separator Clot Activator tubes are evacuated blood collection tubes used for serum determinations. They contain clot-activating micronized silica particles and a gel separator that, once centrifuged, forms a stable barrier separating the serum from fibrin and cells.

  • VACUETTE™ Blood Collection Tubes are evacuated for easy, safe sample collection. The amount of vacuum in each tube corresponds to the tube’s blood volume specification and helps ensure the correct blood-to-additive ratio.
  • Injection molded from polyethylene terephthalate (PET) plastic, VACUETTE™ Blood Collection Tubes are highly transparent and virtually unbreakable.
  • VACUETTE™ Serum tubes include a clot-activating additive – micronized silica particles – that is spray-dried on the interior tube wall for even disbursement and optimal blood/additive mixing.
  • Serum tubes with gel contain a barrier gel to separate the serum from the fibrin and cells. During centrifugation, the barrier gel moves upward to the serum/clot interface, where it forms a stable barrier. This barrier separates the serum from fibrin and cells.
  • Optimal fill marks on every tube label indicate the proper draw volume for correct blood-to-additive ratios (within ±10% of the stated nominal volume in accordance with ISO and CLSI).
  • Featuring red VACUETTE™ SAFETY Caps, designed to minimize the spraying of aerosols during decapping and color-coded to aid first-glance tube recognition.
  • Sterile and latex-free.

Specifications

Color Transparent
Product Type Blood Collection Tube
Additive Clot Activator and Gel
Material PET
Sterility Sterile
Closure Type Non-ridged Pull Cap
Capacity (Metric) 2.5 mL
Diameter (Metric) 13 mm
Closure Color Red with White Ring
Tube Size 13 x 75 mm
Length (Metric) 75 mm
Show More Show Less