missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One VACUETTE™ CAT Serum Separator Clot Activator Blood Collection Tubes
Description
- VACUETTE™ Blood Collection Tubes are evacuated for easy, safe sample collection. The amount of vacuum in each tube corresponds to the tube’s blood volume specification and helps ensure the correct blood-to-additive ratio.
- Injection molded from polyethylene terephthalate (PET) plastic, VACUETTE™ Blood Collection Tubes are highly transparent and virtually unbreakable.
- VACUETTE™ Serum tubes include a clot-activating additive – micronized silica particles – that is spray-dried on the interior tube wall for even disbursement and optimal blood/additive mixing.
- Serum tubes with gel contain a barrier gel to separate the serum from the fibrin and cells. During centrifugation, the barrier gel moves upward to the serum/clot interface, where it forms a stable barrier. This barrier separates the serum from fibrin and cells.
- Optimal fill marks on every tube label indicate the proper draw volume for correct blood-to-additive ratios (within ±10% of the stated nominal volume in accordance with ISO and CLSI).
- Featuring red VACUETTE™ SAFETY Caps, designed to minimize the spraying of aerosols during decapping and color-coded to aid first-glance tube recognition.
- Sterile and latex-free.
Specifications
Specifications
| Color | Transparent |
| Product Type | Blood Collection Tube |
| Additive | Clot Activator and Gel |
| Material | PET |
| Sterility | Sterile |
| Closure Type | Non-ridged Pull Cap |
| Capacity (Metric) | 3.5 mL |
| Diameter (Metric) | 13 mm |
| Closure Color | Red with Yellow Ring |
| Tube Size | 13 x 75 mm |
| Show More |