missing translation for 'onlineSavingsMsg'
Learn More

Serotonin Creatinine Sulfate Monohydrate 98.0+%, TCI America™

Catalog No. S00361G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00361G 1 g
S0036100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S00361G Supplier TCI America Supplier No. S00361G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61-47-2
Molecular Formula C14H23N5O7S
Molecular Weight (g/mol) 405.426
MDL Number MFCD00149653
InChI Key BKCXVJIGPVULPX-UHFFFAOYSA-N
Synonym serotonin creatinine sulfate monohydrate, serotonin creatinin-sulfat-1-wasser, unii-6l522laq9u, serotonin creatine sulfate monohydrate, creatinina serotonin sulfuric acid hydrate, 1-benzo b thien-3-yl-3-1-pyrrolidinyl-1-propanone hydrochloride, 1-propanone, 1-benzo b thien-3-yl-3-1-pyrrolidinyl-, hydrochloride, sulfuric acid compound with 2-imino-1-methylimidazolidin-4-one and 3-2-aminoethyl-1h-indol-5-ol 1:1:1 hydrate, ccris 8960, serotonincreatininesulfatemonohydrate
PubChem CID 164531
IUPAC Name 3-(2-aminoethyl)-1H-indol-5-ol;2-amino-3-methyl-4H-imidazol-5-one;sulfuric acid;hydrate
SMILES CN1CC(=O)N=C1N.C1=CC2=C(C=C1O)C(=CN2)CCN.O.OS(=O)(=O)O

Specifications

Melting Point 215°C
Color White-Yellow
Quantity 1 g
Formula Weight 387.41
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Serotonin Creatinine Sulfate Monohydrate
RTECSNumber NM2550000
TSCA No