missing translation for 'onlineSavingsMsg'
Learn More

Serotonin creatinine sulfate monohydrate, 99%, Thermo Scientific™

Chemical Identifiers

CAS 61-47-2
Molecular Formula H2SO4·H2O
Molecular Weight (g/mol) 405.42
MDL Number MFCD00149653
InChI Key BKCXVJIGPVULPX-UHFFFAOYSA-N
Synonym serotonin creatinine sulfate monohydrate, serotonin creatinin-sulfat-1-wasser, unii-6l522laq9u, serotonin creatine sulfate monohydrate, creatinina serotonin sulfuric acid hydrate, 1-benzo b thien-3-yl-3-1-pyrrolidinyl-1-propanone hydrochloride, 1-propanone, 1-benzo b thien-3-yl-3-1-pyrrolidinyl-, hydrochloride, sulfuric acid compound with 2-imino-1-methylimidazolidin-4-one and 3-2-aminoethyl-1h-indol-5-ol 1:1:1 hydrate, ccris 8960, serotonincreatininesulfatemonohydrate
PubChem CID 164531
IUPAC Name 3-(2-aminoethyl)-1H-indol-5-ol;2-amino-3-methyl-4H-imidazol-5-one;sulfuric acid;hydrate
SMILES CN1CC(=O)N=C1N.C1=CC2=C(C=C1O)C(=CN2)CCN.O.OS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC132680010
SDS View Documents
Thermo Scientific Chemicals
132680010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC132680050
SDS View Documents
Thermo Scientific Chemicals
132680050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

61-47-2
405.42
BKCXVJIGPVULPX-UHFFFAOYSA-N
164531
CN1CC(=O)N=C1N.C1=CC2=C(C=C1O)C(=CN2)CCN.O.OS(=O)(=O)O
H2SO4·H2O
MFCD00149653
serotonin creatinine sulfate monohydrate, serotonin creatinin-sulfat-1-wasser, unii-6l522laq9u, serotonin creatine sulfate monohydrate, creatinina serotonin sulfuric acid hydrate, 1-benzo b thien-3-yl-3-1-pyrrolidinyl-1-propanone hydrochloride, 1-propanone, 1-benzo b thien-3-yl-3-1-pyrrolidinyl-, hydrochloride, sulfuric acid compound with 2-imino-1-methylimidazolidin-4-one and 3-2-aminoethyl-1h-indol-5-ol 1:1:1 hydrate, ccris 8960, serotonincreatininesulfatemonohydrate
3-(2-aminoethyl)-1H-indol-5-ol;2-amino-3-methyl-4H-imidazol-5-one;sulfuric acid;hydrate

Specifications

61-47-2
99%
H2SO4·H2O
1g
serotonin creatinine sulfate monohydrate, serotonin creatinin-sulfat-1-wasser, unii-6l522laq9u, serotonin creatine sulfate monohydrate, creatinina serotonin sulfuric acid hydrate, 1-benzo b thien-3-yl-3-1-pyrrolidinyl-1-propanone hydrochloride, 1-propanone, 1-benzo b thien-3-yl-3-1-pyrrolidinyl-, hydrochloride, sulfuric acid compound with 2-imino-1-methylimidazolidin-4-one and 3-2-aminoethyl-1h-indol-5-ol 1:1:1 hydrate, ccris 8960, serotonincreatininesulfatemonohydrate
BKCXVJIGPVULPX-UHFFFAOYSA-N
3-(2-aminoethyl)-1H-indol-5-ol;2-amino-3-methyl-4H-imidazol-5-one;sulfuric acid;hydrate
164531
99%
Authentic
Glass bottle
MFCD00149653
14, 8462
Solubility in water: freely soluble in water
CN1CC(=O)N=C1N.C1=CC2=C(C=C1O)C(=CN2)CCN.O.OS(=O)(=O)O
405.42
405.42
Serotonin creatinine sulfate monohydrate, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause damage to organs.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only