Learn More
BeLLCo Glass Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Plain top promotes rapid & even flow.
Catalog No. 501944597
Encompass
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Plain top promotes rapid & even flow.
Catalog No. 501944597 Supplier BeLLCo Glass Supplier No. 121210010
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Plain top promotes rapid & even flow.