Learn More
BeLLCo Glass Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Constricted top for cotton plugging. Supp
Catalog No. 501944592
Encompass
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Constricted top for cotton plugging. Supp
Catalog No. 501944592 Supplier BeLLCo Glass Supplier No. 121025010
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Constricted top for cotton plugging. Supp