missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Series 815 Laser Slotted Cassettes with Attached Lids
Description
- Epredia™ Laser Cassettes are designed for use on both the NOVA™ and VEGA™ Laser Printers
- Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
- Each style comes in multiple different colors
- Each cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
- Series 815 come in taped stacks of 40 cassettes with lids attached; 45 stacks per case for a total of 1800 cassettes
- The Epredia™ Laser Cassette is not compatible with thermal tape style printers
- Tape strips on the stacks is made from 95% recycled polypropylene and is entirely recyclable
Specifications
Specifications
| Pore Style | Slots |
| Closure Included | Yes |
| Quantity | 40 Cassettes/Taped Stack, 45 Stacks/Cs. |
| Closure Type | Attached Lid |
| Color | White |
| Product Type | Cassette |