missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Series 315 Laser Embedding Cassettes

Catalog No. 3151218LMI
Change view
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lilac
Ochre
Orange
Pink
Red
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
3151218LMI Red
3150118LMI White
3150418LMI Pink
3150518LMI Green
3150618LMI Orange
3150718LMI Gray
3150818LMI Aqua
3150918LMI Lilac
3151018LMI Beige
3151118LMI Ochre
3150218LMI Yellow
3150318LMI Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 3151218LMI Supplier Epredia™ Supplier No. 3151218LMI

Manufactured specifically for laser printing, the Epredia™ Series 315 Embedding Cassettes have round holes to maximize fluid exchange.

  • Epredia™ Laser Cassettes designed for use on both the NOVA™ and VEGA™ Laser Printers
  • Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Embedding style of cassette comes in different colors
  • Required reusable metal cassette lids (Part number FTML315) are sold separately
  • Embedding cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
  • Series 315 comes in taped stacks of 40 cassettes; 25 stacks per case
  • Epredia™ Laser Cassette Series is not compatible with thermal tape style printers
  • Tape strips on the stacks is made from 95% recycled polypropylene and is entirely recyclable

Specifications

Pore Style Round Holes
Quantity 40 Cassettes/Taped Stack, 45 Stacks/Cs.
Closure Included No
Color Red
Product Type Cassette