missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Series 315 Laser Embedding Cassettes
Description
- Epredia™ Laser Cassettes designed for use on both the NOVA™ and VEGA™ Laser Printers
- Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
- Embedding style of cassette comes in different colors
- Required reusable metal cassette lids (Part number FTML315) are sold separately
- Embedding cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
- Series 315 comes in taped stacks of 40 cassettes; 25 stacks per case
- Epredia™ Laser Cassette Series is not compatible with thermal tape style printers
- Tape strips on the stacks is made from 95% recycled polypropylene and is entirely recyclable
Specifications
Specifications
| Pore Style | Round Holes |
| Quantity | 40 Cassettes/Taped Stack, 45 Stacks/Cs. |
| Closure Included | No |
| Color | White |
| Product Type | Cassette |