missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Series 215 Laser Biopsy Cassettes with Lids

Catalog No. 2151210LMI
Change view
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lilac
Ochre
Orange
Pink
Red
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
2151210LMI Red
2150710LMI Gray
2150810LMI Aqua
2150910LMI Lilac
2151010LMI Beige
2151110LMI Ochre
2150110LMI White
2150210LMI Yellow
2150310LMI Blue
2150410LMI Pink
2150510LMI Green
2150610LMI Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 2151210LMI Supplier Epredia™ Supplier No. 2151210LMI

Manufactured specifically for laser printing, the Epredia™ Series 215 Biopsy Cassettes come with separate lids and are designed for small tissue samples.

  • Epredia™ Laser Cassettes are designed for use on both the Epredia™ NOVA™ and VEGA™ Laser Printers
  • Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Comes in multiple different colors with color-matched lids
  • Designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
  • Comes in taped stacks of 40 cassettes; 25 stacks per case for a total of 1000 cassettes with separate lids
  • Not compatible with thermal tape style printers
  • Tape strips on the stacks is made from 95% recycled polypropylene and is entirely recyclable

Specifications

Pore Style Small Squares
Quantity 40 Cassettes/Taped Stack, 25 Stacks/Cs.
Closure Included Yes
Closure Type Separate Lid
Color Red
Product Type Cassette