missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Series 210 Laser Biopsy Cassettes with Front Tab Lids
Description
- Epredia™ Laser Cassettes are designed for use on both the Epredia™ NOVA™ and VEGA™ Laser Printers
- Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
- Comes in multiple different colors with color-matched lids
- Designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
- Comes in taped stacks of 40 cassettes; 25 stacks per case for a total of 1000 cassettes with separate lids
- Not compatible with thermal tape style printers
- Tape strips on the stacks is made from 95% recycled polypropylene and is entirely recyclable
Specifications
Specifications
| Pore Style | Small Squares |
| Closure Included | Yes |
| Quantity | 40 Cassettes/Taped Stack, 25 Stacks/Cs. |
| Closure Type | Separate Lid with Tab |
| Color | Red |
| Product Type | Cassette |