missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Series 210 Laser Biopsy Cassettes with Front Tab Lids

Catalog No. 2100410LMI
Change view
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lilac
Ochre
Orange
Pink
Red
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
2100410LMI Pink
2100610LMI Orange
2100710LMI Gray
2100810LMI Aqua
2100110LMI White
2100910LMI Lilac
2100210LMI Yellow
2101010LMI Beige
2101110LMI Ochre
2101210LMI Red
2100510LMI Green
2100310LMI Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 2100410LMI Supplier Epredia™ Supplier No. 2100410LMI

Manufactured specifically for laser printing, the Series 210 Biopsy Cassettes come with separate lids with a tab in the front for easy opening and closing.

  • Epredia™ Laser Cassettes are designed for use on both the Epredia™ NOVA™ and VEGA™ Laser Printers
  • Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Comes in multiple different colors with color-matched lids
  • Designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
  • Comes in taped stacks of 40 cassettes; 25 stacks per case for a total of 1000 cassettes with separate lids
  • Not compatible with thermal tape style printers
  • Tape strips on the stacks is made from 95% recycled polypropylene and is entirely recyclable

Specifications

Pore Style Small Squares
Closure Included Yes
Quantity 40 Cassettes/Taped Stack, 25 Stacks/Cs.
Closure Type Separate Lid with Tab
Color Pink
Product Type Cassette