missing translation for 'onlineSavingsMsg'
Learn More

Seratrodast 98.0+%, TCI America™

Catalog No. p-7135999
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0943100MG 100 mg
S09431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0943100MG Supplier TCI America Supplier No. S0943100MG
Only null left

Chemical Identifiers

CAS 112665-43-7
Molecular Formula C22H26O4
Molecular Weight (g/mol) 354.446
MDL Number MFCD00875701
InChI Key ZBVKEHDGYSLCCC-UHFFFAOYSA-N
Synonym seratrodast, bronica, abbott 73001, abbott-73001, seratrodast usan:inn, 7-phenyl-7-2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl heptanoic acid, bronica tn, ccris 8939, +--7-3,5,6-trimethyl-1,4-benzoquinon-2-yl-7-phenylheptanoic acid, seratrodast jan/usan/inn
PubChem CID 2449
IUPAC Name 7-phenyl-7-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)heptanoic acid
SMILES CC1=C(C(=O)C(=C(C1=O)C)C(CCCCCC(=O)O)C2=CC=CC=C2)C

Specifications

Melting Point 128°C
Quantity 100 mg
Formula Weight 354.45
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Seratrodast
RTECSNumber DA1600050
TSCA No
Recommended Storage Freezer