missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag™ Streptavidin Magnetic Beads, low biotin binding capacity

Catalog No. 09981126
Change view
Click to view available options
Quantity:
1 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09981126 1 mL
09981128 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09981126 Supplier Cytiva Supplier No. 30152103011150

Their cauliflower-like structure gives Sera-Mag beads high binding capacity. Sera-Mag Streptavidin beads contain covalently bound streptavidin and are available with low, medium, or high nominal biotin binding capacities. Choose the biotin-binding capacity needed to optimize your application.

Biotin Binding Capacity of 2500 to 3500pmol/mg beads

Specifications

Concentration 1% solids (10mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets
Quantity 1 mL
Diameter (Metric) 1 μm