missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag™ Streptavidin Magnetic Beads, high biotin binding capacity

Catalog No. 09981134
Change view
Click to view available options
Quantity:
1 mL
5 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09981134 100 mL
09981133 5 mL
09981132 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09981134 Supplier Cytiva Supplier No. 30152105010350
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Their cauliflower-like structure gives Sera-Mag beads high binding capacity. Sera-Mag Streptavidin beads contain covalently bound streptavidin and are available with low, medium, or high nominal biotin binding capacities. Choose the biotin-binding capacity needed to optimize your application.

Biotin Binding Capacity of 4500 to 5500pmol/mg beads

Specifications

Concentration 1% solids (10mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets
Quantity 100 mL
Diameter (Metric) 1 μm