missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag SpeedBeads Streptavidin-Blocked Magnetic Particles

Catalog No. 09981912
Change view
Click to view available options
Packaging:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
09981912 5 mL
09981911 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09981912 Supplier Cytiva Supplier No. 21152104010150
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Combines high affinity for biotinylated target molecules with very low nonspecific binding.

  • Combines fast reaction kinetics with very low non-specific binding for increased throughput and precision
  • Applications include automated immunoassays, immunoprecipitation, and protein purification
  • Streptavidin coating targets Biotinylated Molecules
  • Compounds that are difficult to attach to particle surfaces by conventional means may be amenable to biotinylation
  • Provides a high biotin-binding capacity along with a strong affinity for targeted, biotin labeled molecules
  • The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer
  • Two layers of magnetite are coated onto this core, resulting in faster magnetic response times
  • The surface is blocked with a non-surfactant, non-protein based method, to help prevent non-specific binding of proteins
  • Specifications

    Concentration 10 mg/mL
    Packaging 5 mL
    Diameter (Metric) 1 μm