missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag SpeedBeads™ Protein A/G Magnetic Beads

Catalog No. 09981922
Change view
Click to view available options
Quantity:
1 mL
5 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09981922 100 mL
09981921 5 mL
09981920 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09981922 Supplier Cytiva Supplier No. 17152104010350
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provide a fast, convenient method for manual and automated magnetic isolation of IgA and IgG proteins using affinity binding. The beads can be used with the broadest range of antibody species and subclasses (even when subclass identity has not been determined).

  • Their 'cauliflower-like' structure gives Sera-Mag beads high binding capacity
  • Fast and easy IgA purification and IgG purification

Specifications

Concentration 1% solids (10mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) Affinity Purification
Quantity 100 mL
Diameter (Metric) 1 μm