missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag SpeedBeads™ Carboxyl Magnetic Beads, hydrophilic
Excellent for coupling oligos or proteins directly to the beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.
Supplier: Cytiva 45152105050350
Description
- Double shell magnetite beads
- . High binding capacity
- 1.0μm diameter
- High carboxyl acid content for binding for binding DNA and proteins
- Hydrophilic bead based on crosslinking requirements for the encapsulation layer
Chemical Identifiers
| 5% solids (50mg/mL) | |
| 2°C to 8°C | |
| DNA Sample Prep, Protein Purification | |
| 1 μm |
Specifications
| 5% solids (50mg/mL) | |
| DNA Sample Prep, Protein Purification | |
| 1 μm |
| 2°C to 8°C | |
| 100 mL |