missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag™ Oligo d(T) Magnetic Beads

Catalog No. 09981145
Change view
Click to view available options
Quantity:
1 mL
5 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09981145 5 mL
09880012 1 mL
09880014 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09981145 Supplier Cytiva Supplier No. 38152103010150
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suited for capturing or isolating mRNA from a variety of sources. Due to their cauliflower like structure, Sera-Mag beads have high binding capacity. Oligo dT magnetic beads contain covalently bound oligo(dT)14.

  • Colloidally stable and will remain in suspension for extended periods of time in the absence of a magnetic field
  • High binding capacity
  • 1.0μm diameter
  • High oligo dT content for mRNA binding

Specifications

Concentration 1% solids (10mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) mRNA Purification
Quantity 5 mL
Diameter (Metric) 1 μm