missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Sequence Detection Systems Spectral Calibration Kit, 384-well

Catalog No. 4323977
Encompass
Change view
Click to view available options
Quantity:
2 x 384-well Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4323977 2 x 384-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4323977 Supplier Applied Biosystems™ Supplier No. 4323977
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Easy-to-use, preloaded, 96-well optical reaction plates enable users to calibrate the 384-well block of the 7900HT Sequence Detection System quickly and easily.

This Applied Biosystems™ easy-to-use kit establishes the pure dye spectra and multicomponent values for FAM™, JOE™, NED™,ROX™, SYBR™ Green, TAMRA™, TET™, and VIC™ dyes on the 7900HT Sequence Detection System. The Sequence Detection Systems Spectral 384-well Calibration kit includes two optical 384-well reaction plates: one preloaded and sealed background plate, and one preloaded and sealed spectral calibration plate with eight separate dye standards. Spend Your Time on Research - Not Instrument Calibration This kit's easy-to-use, preloaded, 96-well optical reaction plates enable users to calibrate the 384-well block of the 7900HT Sequence Detection System quickly and easily. Insert the reaction plates during system installation, and the system automatically collects the dye standard's spectral information. The system's application algorithm then uses the calibration information to ensure accurate data analysis.

  • 96-well or 384-well optical reaction plates preloaded with dyes (FAM™, JOE™, NED™, ROX™, SYBR™ Green, TAMRA™, TET™, and VIC™) are provided for quick and easy calibration of the ABI PRISM™ 7900HT Sequence Detection System
  • Easy-to-use and provides accurate discrimination of the fluorescence signal of individual dyes in the reaction mixture

For Research Use Only. Not for use in diagnostic procedures.

Order Info

Expiration date: 12 months from the date of delivery.

Specifications

Content And Storage Includes one Background and one Spectral Calibration plate with eight dye standards plates (FAM™, JOE™, NED™, ROX™, SYBR™ Green, TAMRA™, TET™, and VIC™).

Store at -15 to 30°C. Protect from light.

Expiration date: 12 months from the date of delivery
Calibrated Dye FAM, JOE, NED, ROX, TAMRA™, TMR (Tetramethylrhodamine), VIC, SYBR™ Green
For Use With (Equipment) AB 7900 HT 384-well System
Detection Method Fluorescence
Product Type Spectral Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Description 384-well Plate
Quantity 2 x 384-well Plates
Calibration Type Spectral Calibration
How can I convert a file from AQ to RQ, or from RQ to AQ file with the 7900HT Fast Real-Time PCR System?

SDS v2.4 and above allow you to convert your AQ files to RQ and vice versa.
To do so, simply go to: Tools then AQ RQ Converter
-Choose from the dropdown which direction you want to go (AQ to RQ, or RQ to AQ).
-Next click on ‘Add' and browse to the plate file location in the new window. and click ‘Open'.
-Click on ‘Convert'. A new file will be created in the same location as the original, but with _AQ or _RQ at the end of the filename.

If you do not have v2.4, you can upgrade your software here (http://www.thermofisher.com/us/en/home/technical-resources/software-downloads/applied-biosystems-7900ht-fast-real-timespcr-system.html).

One benefit of this conversion feature is that it allows you to convert 7900 files to the correct type to do analysis for relative quantification study, for example, if a plate is originally run as AQ instead of RQ, or if you would like to generate a standard curve quantification in the AQ if a plate was originally run as RQ.

Why are there no RQ values for all the samples in my study in the 7900HT Fast Real-Time PCR System software?

The 7900HT Fast Real-Time PCR System software requires that the endogenous control be run on every plate within a study. If not, you will only see RQ values for the plate that contains the endogenous control on it. The other plates will have Ct values for all the wells, but the software will not calculate RQ values. If you designed your experiments in this type of layout, you will want to do your analysis in the free DataAssist Software (https://www.thermofisher.com/us/en/home/technical-resources/software-downloads/dataassist-software.html?icid=fr-realtimesoftware-5) instead, or use the Thermo Fisher Cloud (https://www.thermofisher.com/us/en/home/cloud.html). This program does not have the endogenous control requirement for every plate. Simply export the study results file from the 7900HT software as a *.txt or *.csv file, and use that to create a new study in the DataAssist software. You should now be able to see RQ values for all samples, even those without an endogenous control on the same plate.

Why do I not see any Ct or RQ values from my ddCT experiment in the 7900HT Fast Real-Time PCR System software?

In the 7900HT Fast Real-Time PCR System software, v2.3 or later, CT or RQ values are only provided in the RQ Manager software. RQ Manager is installed on the computer along with SDS v2.x.

What can I use for data analysis on the 7900HT Fast Real-Time PCR System?

For genotyping data, we recommend TaqMan Genotyper Software (https://www.thermofisher.com/us/en/home/technical-resources/software-downloads/taqman-genotyper-software.html?icid=fr-software-1).
For relative quantitation, we recommend ExpressionSuite Software (https://www.thermofisher.com/us/en/home/technical-resources/software-downloads/expressionsuite-software.html). You may also use the Thermo Fisher Cloud for either application. https://www.thermofisher.com/us/en/home/cloud.html

What dyes can I use on the 7900HT Fast Real-Time PCR System?

The Applied Biosystems 7900HT Fast Real-Time PCR Systems use the following dye sets for calibration: FAM, SYBR, VIC, ROX, NED, TAMRA, JOE, TET dyes. Custom dyes can also be used that read between 500 nm and 660 nm, although you will have to calibrate the system first for any new dye.

How do I add detectors to a plate document on the 7900HT Fast Real-Time PCR System?

Detectors can be added to a plate before or after a run is complete. To do so:

-Go to Tools then Detector Manager.
-Find the Detector that you want to add from the list, and select it by clicking on it.
-Click on ‘Copy to Plate Document'. (If you have more than one detector to add, repeat these steps.)
-When finished, click “Done”. You should now see the Detectors you added under the Setup Tab.
-Now highlight the wells that you want and check the box next to the detector you want to use. Repeat for however many detectors you need. Analyze the plate by clicking on the Green Arrow and then ‘Save As' a new file name.

How do I add a melt curve to my experiment on the 7900HT Fast Real-Time PCR System?

In the 7900HT SDS software, you can run a melt curve at the end of an Absolute Quantification plate run, or run a melt curve by itself. You cannot directly add a melt curve to the end of a Relative Quantification (or ddCT plate).

For the former, here are the steps:
-File then New, Assay: Standard Curve (AQ), Click ‘OK'.
-In the new document, click on the ‘Instrument' tab (right hand side). You should now see the thermal profile.
-Click in the box at the last step such that a black line appears. Click on the ‘Add Dissociation Stage' button which should now be active.

If you just want to run a melt curve (no amplification stage), then you will want to highlight the other steps (click and drag across Stages 1-3). Then click on the ‘Delete Step' button. Now you can run just the melt curve.If you want to run a melt curve with a RQ file, you have two options. One, if you have the v2.4 software, you can run the file as demonstrated above as AQ with a melt curve, and then use the conversion tool later to switch to an RQ file. The other option is to run the file as RQ, and then once the run is complete, set up the melt curve run as described above.

Can I use the 7900HT Fast Real-Time PCR System to do a protein thermal melt experiment?

Yes, but the data is not compatible with our Protein Thermal Shift software. Below are instructions on how to program the software and a reference for the analysis.

1.Create a new experiment using File then New in the 7900HT software. Click ‘OK'.
2.Go to the bottom of the Setup panel, select Passive Reference as None.
3.Click on ‘Add Detector' button on the setup panel.
-If you do not have a preset detector,click on ‘New' from Detector Manager window, so you can define a new detector. Make sure the reporter dye is ROX, and quencher is Non Fluorescent.
-Once you have the desired detector highlighted in the Detector Manager window, click the ‘Copy to Plate Document' button, then click ‘Done'. You will see the detector now in the Setup panel.
4.Next go to Instrument tab, in which you will see the default the thermal profile.
5.Click at the end of the thermal profile, a vertical line will appear.
6.Click the ‘Add Dissociation Stage' button, then highlight and delete the PCR stages.
7.Edit the profile as below, to go from 25 degrees C (2 min) to 95 degrees C (15 sec).
8.Click on the ‘Ramp Rate' tab, and change the ramping to 1%.
9.Save the file, and start the run (or save as a template).
Please note that you will have to perform the data analysis separately from the raw data. You can refer to this reference as a starting point (Nat Protoc 2007;2(9):2212-21).

Where are my files saved to?

On the 7900HT Fast Real-Time PCR System, run files will be saved to a default folder on the connected computer, unless you change it.
To find or change the default folder, go to Tools then Options then General. Here you will see a Data Folder and an Import Folder. The default location is shown. If you want files to be saved to (or open from) a different location, click ‘Browse' and choose the new folder.

Can I collect data at two separate steps for analysis?

No. On the 7900HT Fast Real-Time PCR System, the software will use the last data collection step in the cycling stage for all amplification plots and Ct analysis. So even if you were to set two separate steps with ‘Data Collection On' you would only be able to view and analyze data from the latter step.

What volumes can be used in my plate on the 7900HT Fast Real-Time PCR System?

The following volumes are supported for each 7900HT instrument block:

-Standard 96-well block: 20-50 µL
-Fast 96-well block: 10-30 µL
-384-well block: 5-20 µL
-TaqMan Low Density Array block: approximately 1µL

Can I use the 7900HT Fast Real-Time PCR System instrument computer while a run is in progress?

Using the instrument computer when a run is in progress is not recommended, as this poses a risk of corrupting the data.

How do I add a custom dye to the 7900HT Fast Real-Time PCR System?

The 7900HT Fast Real-Time PCR System can be used to run assays designed with custom dyes (dyes not manufactured by Life Technologies). Custom dyes must fluoresce within the 500-660 nm spectral range measured by the 7900HT instrument. The calibration is done in two steps. In the first step, you will need to obtain an oligo labeled with your dye (no quencher), and then create a plate containing different concentrations of the oligo (approximately 25-3,200 nM). This is done to determine the optimal concentration to use for the actual calibration. In the second step, you will create a plate of the oligo at the concentration determined in the previous step and then run a custom dye calibration. You need to fill at least 3 full columns for adequate data collection. Follow the directions in Appendix B of the 7900HT Fast Real-Time PCR System Maintenance Guide (http://tools.thermofisher.com/content/sfs/manuals/cms_042134.pdf) for more specific instructions.

How do I use tubes in the 7900HT Fast Real-Time PCR System?

Do not use MicroAmp Optical Caps or MicroAmp Optical Tubes with the 7900HT Fast Real-Time PCR System. The instrument is not designed to run MicroAmp Tubes. Please be aware that use of tubes may damage its internal components.

What are the specifications for the 7900HT Fast Real-Time PCR System?

Instrument Specifications
Block Options (Interchangeable)
96-well (standard), 96-well (Fast), 384-well, TaqMan Low Density Array
Sensitivity Down to 1 copy (2-fold discrimination with 99.7% confidence)
Dynamic Range 9 logs of linear dynamic range
Calibrated Dyes FAM, SYBR, VIC, ROX, NED, TAMRA, JOE, TET dyes
Detection Method SYBR dye, primer-probe detection
Resolution Detect changes as little as 1.5-fold
Reaction Volume Range 20-50 µL Standard 96-well, 10-30 µL Fast 96-well block, 5-20 µL 384-well, ~ 1 µL 384-well TaqMan Low Density Array
Reaction Speed 9600 emulation mode, Standard, and Fast
Optics Extended-life 488 nm argon-ion laser excitation source
No filters (CCD acts as spectrograph with continuous detection 500-650 nm)

Temperature Range 4-100 degrees C
Run Time <2 hr (standard mode)
Approximately 35 min (Fast mode)

Temperature Accuracy plus or minus 0.25 degrees C (between 35 degrees C and 95 degrees C, after 3 min)
Temperature Uniformity plus or minu 0.5 degrees C (after 30 sec)
Thermal Cycling System Peltier-based system
Available Applications Gene expression, genotyping, copy number variation, HRM, protein thermal shift, protein detection, mutation detection, miRNA, presence/absence
Dimensions 72 cm (W) x 84 cm (D) x 64 cm (H) (with drawer open)
Weight 82 kg (180 lb) without automation accessory
114 kg (250 lb) with automation accessory
Remote Monitoring No
On-Board Memory No
Setup Configurations PC-controlled only

What do I do if the background calibration fails on the 7900HT Fast Real-Time PCR System?

Follow the directions in the 7900HT Fast Real-Time PCR System Maintenance Guide, Chapter 7 (http://tools.thermofisher.com/content/sfs/manuals/cms_042134.pdf), to decontaminate the sample block.

When do I need to run the RNaseP calibration on the 7900HT Fast Real-Time PCR System?

The RNaseP verification plate contains template, master mix, and a TaqMan assay for RNaseP. It is used to verify that the instrument is performing to specifications. If you have reason to suspect there is something wrong with the instrument, or if you want to rule out a chemistry issue, the RNaseP plate is a good way to test the system. The RNaseP verification plate is a single-use plate.

Can I reuse my calibration plates on the 7900HT Fast Real-Time PCR System?

The calibration plates can be stored and reused three times for up to 12 months after you first open them, so make sure to return them to their original packaging and return them to -20 degrees C storage until the next use. (Note: Due to the small volumes, calibration array cards cannot be reused. Please discard them after use.) If needed, you can make your own background plate using deionized water.

When do I need to calibrate the 7900HT Fast Real-Time PCR System?

Recommended Maintenance Schedule

Power on/off the computer controlling the instrument: Weekly
Check computer disk space. If necessary, archive or back up your experiment files and instrument settings: Weekly
Run disk cleanup and disk defragmentation: Every month
Background calibration: Every month
Perform an instrument self test: Every month
Pure dye calibrations: Every 6 months
RNaseP instrument verification: After the instrument has been moved, or as needed to verify instrument performance

What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.


For Research Use Only. Not for use in diagnostic procedures.