missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SeqStudio™ Integrated Capillary Protector

Catalog No. A31923
Encompass
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31923 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A31923 Supplier Applied Biosystems™ Supplier No. A31923
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems SeqStudio Integrated Capillary Protector (ICP) is used on the SeqStudio Cartridge. Gel-filled and is for one-time-use only.

The Applied Biosystems™ SeqStudio™ Integrated Capillary Protector (ICP) is used on the SeqStudio Cartridge. The Integrated Capillary Protector protects the capillary ends on the cartridge. The protector is gel-filled and is for one-time-use only.

Order Info

Each

Specifications

Content And Storage Store in refrigerator (2–8°C).
Product Line SeqStudio
Type Integrated Capillary Protector
Quantity 1 each
Shipping Condition Approved for shipment at room temperature or wet ice
What is included in the SeqStudio Cartridge v2 (Cat. No. A41331) and can the cartridge (Cat. No. A41328) be purchased separately?

The SeqStudio Cartridge v2 includes the SeqStudio Genetic Analyzer Cartridge v2 and a SeqStudio Integrated Capillary Protector (Cat. No. A31923). Even though the cartridge has "A41328" on the box, it cannot be purchased separately and is only available as part of the SeqStudio Cartridge v2.

What does the SeqStudio Starter Kit (PN A35000) contain?

The SeqStudio Starter Kit (PN A35000) includes:
- SeqStudio Integrated Capillary Protector (Cat. No. A31923)
- SeqStudio Cathode Buffer Container (Cat. No. A33401)
- Sequencing Standard, BigDye Terminator v3.1, for 3500/SeqStudio Flex (Cat. No. 4404312)
- DS-33 GeneScan Installation Standards with GeneScan 600 LIZ Size Standard v2.0 (Cat. No. 4376911)
- Hi-Di Formamide (Cat. No. 4401457)
- MicroAmp Optical 96-Well Reaction Plate with Barcode (Cat. No. 4306737)
- MicroAmp 96-Well Tray/Retainer Set (Cat. No. 403081)
- MicroAmp Optical 8-Tube Strip, 0.2 mL (Cat. No. 4316567)
- Nuclease-Free Water (not DEPC-Treated) (Cat. No. AM9930)
- Cathode Buffer Container Reservoir Septa for SeqStudio Genetic Analyzer (Cat. No. A35640)
- 96-well Septa for SeqStudio Genetic Analyzer (Cat. No. A35641)
- 8-strip Septa for SeqStudio Genetic Analyzer (Cat. No. A35643)
- SeqStudio Cartridge v2 (PN A41328) (if this component is ordered online the catalog number is A41331)

For Research Use Only. Not for use in diagnostic procedures.