missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SeqStudio™ Genetic Analyzer System with SmartStart

Catalog No. A35644
Encompass
Change view
Click to view available options
Content And Storage:
System + SmartStart + 1 yr ext warranty
System + SmartStart + 3 yr ext warranty
System + SmartStart + Software
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
A35644 System + SmartStart + Software
A35645 System + SmartStart + 1 yr ext warranty
A35646 System + SmartStart + 3 yr ext warranty
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A35644 Supplier Applied Biosystems™ Supplier No. A35644

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis system designed to provide users with an integrated experience that is easy, efficient, and versatile for research in genetic analysis.

  • The Applied Biosystems SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis system designed to provide users with an integrated experience that is easy, efficient, and versatile for research in genetic analysis
  • Combining an extended warranty with your system purchase, Help extend the life of your investment. These packages include the system, SmartStart, and an extended warranty.
  • Simplified instrument software with interactive touch screen for greater ease of use
  • Minimized hands-on setup time with integrated reagents cartridge (SeqStudio Cartridge)
  • Application flexibility to run Sanger sequencing and fragment analysis on the same run
  • Connectivity: Wi-Fi or wired Ethernet connection
  • Web-enabled remote monitoring capability
  • Consumables usage tracking through radio frequency identification (RFID)
  • SmartStart is performed by a Field Application Scientist (FAS) in your lab. SmartStart helps you:
  • Get up and running quickly
  • Get connected to your local network and Cloud
  • Choose appropriate reagents and consumables to meet your needs
  • Content will vary depending on your level of knowledge and experience.

Specifications

Content And Storage System + SmartStart + Software
For Use With (Application) Sanger Sequencing
Product Line SeqStudio
Type Genetic Analyzer
Have you tested the SeqStudio Genetic Analyzer (Cat. No. A35644, A35645) in low humidity conditions (below 20%)?

SeqStudio Genetic Analyzer has not been tested for operation in low humidity conditions and is validated for environments with 20-80% humidity. Therefore, we do not recommend using SeqStudio Genetic Analyzer in below 20% humidity.

If the computer attached to the SeqStudio Genetic Analyzer System with SmartStart is upgraded to Win 10, does the data collection software need to be upgraded as well?

No, since the SeqStudio instrument's data collection software is on the instrument itself, it is not necessary that you upgrade it.

Can I run SNaPshot applications, between 35 and 120 bp, on the SeqStudio Genetic Analyzer and the Applied Biosystems 3500 Genetic Analyzer with a 50 cm capillary array?

Both the SeqStudio Genetic Analyzer and the Applied Biosystems 3500 Genetic Analyzer have run modules specific for the SNaPshot application.

- For the 3500 Genetic Analyzer, the run module is "SNaPshot50_POP7" and it has a ~30 minute run time.
- For the SeqStudio Genetic Analyzer, the run module is "SNaPshot", the optimal size range is 40-120 bp, and it runs for ~25 minutes but the 35 bp fragment will be collected.

How do I set up the same user, system security, audit, and e-signature settings on multiple instances of the SAE Administrator Console v2.0?

You may use the export and import functions to transfer settings from one installation of the SAE Administrator Console to another.

What does archiving audit records in the SAE Administrator Console do?

Archiving audit records removes the records from the SAE Administrator Console and saves them in an internally specified location on the same computer on which the SAE Administrator Console is installed. Note: Archived audit records are accessible for viewing in the SAE Administrator Console.

What actions are auditable in the SAE Administrator Console v2.0?

Auditable actions in the SAE Administrator Console are:
- Enable or disable security, audit, or e-signature
- Sign in to or out of the SAE Administrator Console
- Import or export an SAE configuration
- Install an application profile
- Archive, purge, or restore audit records
- Manual sync with LDAP (Lightweight Directory Access Protocol) Directory

Can I enable or disable the system security function when using the SAE Administrator Console v2.0?

The system security function cannot be disabled in the SAE Administrator Console. To disable user sign in to an application, you must disable SAE in the application.

What happens if I forget my password when using the SAE Administrator Console v2.0?

There is no way to recover a forgotten password. If the SAE administrator forgets their password, the software must be reinstalled. Export all data before reinstalling the software.

What actions are audited silently on the SeqStudio instrument when using the SAE Administrator Console v2.0?

The following user actions are audited silently (no audit reason dialog box is displayed), and you cannot change the Audit mode:
- Enable or disable SAE functions in the SeqStudio Data Collection Software
- Sign in to or sign out of the SeqStudio Data Collection Software
- Start, pause, resume, or cancel an instrument injection or plate run
- Perform a dye calibration or an install run
- Perform cartridge maintenance: Refresh PDS or Fill Array

What are the default audit and e-signature settings when SAE is enabled in the SeqStudio Data Collection Software?

When SAE is enabled in the SeqStudio Data Collection Software, the SAE functions are set to the following:
- Audit: Enabled by default, but can be disabled
- E‑signature: Enabled by default, but can be disabled. By default, no actions are set to require e‑signature.

How does the SAE Administrator Console v2.0 affect the way I use the SeqStudio instrument?

When the SAE module is enabled:
- The SAE module controls the access to and the available functions for the SeqStudio Data Collection Software.
- The SAE module does not affect SeqStudio Plate Manager desktop or cloud.
- The Connect cloud-based platform and remote monitoring functions are disabled in the SeqStudio Data Collection Software.

On what operating system can I install the SAE Administrator Console v2.0?

The SAE Administrator Console can be installed on a Windows 7 or Windows 10 operating system.

What are the components of the SAE module?

The SAE module is a client-server software configuration that includes three components:
- SAE Administrator Console: Tool that is used by an SAE administrator to configure the SAE module.
- SAE server (server): Service that runs in the background and stores SAE settings, user accounts, audit records, and e-signature records. By default, the SAE server is installed on the same computer as the SAE Administrator Console.
- SAE screens (client): Screens that are displayed in an application (sign in, audit, and e-signature) and that require user input. The SeqStudio Data Collection Software runs on the client.

What is the SAE Administrator Console v2.0?

The SAE Administrator Console is the tool that is used to configure the Security, Audit, and Electronic Signature (SAE) module. The SAE module can be configured to meet specific requirements for security, audit, and e-signature. In the SAE Administrator Console, a software or instrument that is configured for the SAE module is called an "application". Example applications include the SeqStudio Data Collection Software or the QuantStudio Design and Analysis Software v2.

Where can I find the SAE Administrator Console v2.0 User Guide?

The SAE Administrator Console v2.0 User Guide may be found here: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017468_SeqStudioSAE_UG.pdf


For Research Use Only. Not for use in diagnostic procedures.