missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SeqStudio™ Cathode Buffer Container

Catalog No. A33401
Encompass
Change view
Click to view available options
Quantity:
4 containers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A33401 4 containers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A33401 Supplier Applied Biosystems™ Supplier No. A33401

The Applied Biosystems SeqStudio Cathode Buffer Container (CBC) contains running buffer to support Sanger sequencing and fragment analysis applications on the SeqStudio Genetic Analyzer System.

  • Ready-to-use, disposable container with running buffer
  • Buffer usage tracking with radio frequency identification (RFID)
  • Two week on-instrument shelf life after installation

Order Info

4 containers

Specifications

Content And Storage Store in refrigerator (2–8°C).
Product Line SeqStudio
Type Cathode Buffer Container
Quantity 4 containers
Shipping Condition Approved for shipment on Wet or Dry Ice
What does the SeqStudio Starter Kit (PN A35000) contain?

The SeqStudio Starter Kit (PN A35000) includes:
- SeqStudio Integrated Capillary Protector (Cat. No. A31923)
- SeqStudio Cathode Buffer Container (Cat. No. A33401)
- Sequencing Standard, BigDye Terminator v3.1, for 3500/SeqStudio Flex (Cat. No. 4404312)
- DS-33 GeneScan Installation Standards with GeneScan 600 LIZ Size Standard v2.0 (Cat. No. 4376911)
- Hi-Di Formamide (Cat. No. 4401457)
- MicroAmp Optical 96-Well Reaction Plate with Barcode (Cat. No. 4306737)
- MicroAmp 96-Well Tray/Retainer Set (Cat. No. 403081)
- MicroAmp Optical 8-Tube Strip, 0.2 mL (Cat. No. 4316567)
- Nuclease-Free Water (not DEPC-Treated) (Cat. No. AM9930)
- Cathode Buffer Container Reservoir Septa for SeqStudio Genetic Analyzer (Cat. No. A35640)
- 96-well Septa for SeqStudio Genetic Analyzer (Cat. No. A35641)
- 8-strip Septa for SeqStudio Genetic Analyzer (Cat. No. A35643)
- SeqStudio Cartridge v2 (PN A41328) (if this component is ordered online the catalog number is A41331)

For Research Use Only. Not for use in diagnostic procedures.