missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ SeqStudio™ Cartridge v2
Description
Key features:
- Minimize hands-on setup time with all-in-one cartridge
- Track consumables usage through radio frequency identification (RFID)
- Extended on-instrument 6-month shelf life for cartridge after opening
- Allows for 250 injections or 1000 reactions per cartridge*
- Responsible, simple, no-charge recycling program
* SeqStudio Cartridge v2 is optimized to work with the latest software version for the Seqstudio Genetic Analyzer. Please ensure that Seqstudio Genetic Analyzer is running the latest software before using this cartridge.
Specifications
Specifications
| For Use With (Application) | Comparative Sequencing (Somatic Mutations - 10:50), De Novo Sequencing - Low to Mid Throughput, Fragment Analysis (Denaturing), Rare Variant Sequencing, Resequencing, Short PCR Product Sequencing, Short Read Sequencing |
| For Use With (Equipment) | SeqStudio Genetic Analyzer System |
| Includes | Capillaries, Universal polymer, Buffer, Pump |
| No. of Reactions | 1000 |
| Size | 250 injections/1000 reactions |
| Temperature (Metric) Storage | Store in refrigerator (2 to 8°C) |
| Length (Metric) | Capillary length: 28 cm |