missing translation for 'onlineSavingsMsg'
Learn More

STERIS Septihol™ Sterile Alcohol Solution

A ready to use 70% (v/v) USP-grade sterile isopropyl alcohol solution that kills a range of Gram-positive and Gram-negative bacteria and has fungicidal activity against Trichophyton interdigitale. Double bagged and gamma irradiated for use in aseptic areas.

$401.87 - $725.25

Specifications

Product Type Alcohol Solution
Sterility Sterile
Composition 70% (v/v) USP-Grade Sterile Isopropyl Alcohol
Format RTU
For Use With (Application) Removing Residue From Surfaces, Decontaminating Gloves
View More Specs

Products 2
Catalog Number Mfr. No. Volume (English) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (English) Price Quantity & Availability  
19415111
View Documents Promotion Details
STERIS
6248Y4
16 oz.
Case of 12 Each for $401.87
Only null left
 
19415112
View Documents Promotion Details
STERIS
624896
32 oz.
Case of 12 Each for $725.25
Only null left
 
Description

Description

  • Ideal as a final microbial control and disinfection step to remove residue from surfaces and to decontaminate gloves
  • EPA biocidal and fungicidal claims are proven in the presence of 5% organic soil on hard, non-porous surfaces
  • Can be used straight out of the container, poured or sprayed
  • Ready to use sterile alcohol eliminates in-house production validation and documentation concerns
  • Free of stabilizers and denaturants for quick and complete evaporation
  • Double-bagged and gamma irradiated for use in aseptic areas
  • Each lot is documented with a Certificate of Analysis for lot traceability
Specifications

Specifications

Alcohol Solution
70% (v/v) USP-Grade Sterile Isopropyl Alcohol
Removing Residue From Surfaces, Decontaminating Gloves
Sterile
RTU
Videos
SDS
Documents

Documents