missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STERIS Septihol™ Non-Sterile Alcohol Solution
A 70% (v/v) USP-grade isopropyl alcohol solution that removes residues and has a fast evaporation rate. Ideal for use as a final decontamination step when cleaning.
Supplier: STERIS 625408
Description
- A ready-to-use, 70% USP-grade isopropyl alcohol solution
- Ideal for the removal of process residues and routine glove decontamination
- Free of stabilizers and denaturants for quick and complete evaporation
- Filtered to 0.22 micron
- Available in ergonomically designed, adjustable trigger spray bottles or gallon, bulk bottles
- Can be used straight out of the container, poured or sprayed
- Easily identify non-sterile and sterile versions with different colored triggers
- Each lot is documented with a Certificates of Analysis that includes filtration results
Chemical Identifiers
| Alcohol Solution | |
| 70% (v/v) USP-Grade Isopropyl Alcohol | |
| Removal of Process Residues, Routine Glove Decontamination |
| Non-sterile | |
| Filtered |
Specifications
| Alcohol Solution | |
| 70% (v/v) USP-Grade Isopropyl Alcohol | |
| Filtered | |
| 4/Cs. | |
| 3.8 L |
| Non-sterile | |
| Plastic Bottle | |
| Removal of Process Residues, Routine Glove Decontamination | |
| 1 gal. |