missing translation for 'onlineSavingsMsg'
Learn More

STERIS Septihol™ Non-Sterile Alcohol Solution

Catalog No. 19415114
Change view
Click to view available options
Volume (English):
22 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19415114 22 oz.
19415115 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19415114 Supplier STERIS Supplier No. 625477
Only null left

A 70% (v/v) USP-grade isopropyl alcohol solution that removes residues and has a fast evaporation rate. Ideal for use as a final decontamination step when cleaning.

  • A ready-to-use, 70% USP-grade isopropyl alcohol solution
  • Ideal for the removal of process residues and routine glove decontamination
  • Free of stabilizers and denaturants for quick and complete evaporation
  • Filtered to 0.22 micron
  • Available in ergonomically designed, adjustable trigger spray bottles or gallon, bulk bottles
  • Can be used straight out of the container, poured or sprayed
  • Easily identify non-sterile and sterile versions with different colored triggers
  • Each lot is documented with a Certificates of Analysis that includes filtration results

Specifications

Product Type Alcohol Solution
Sterility Non-sterile
Composition 70% (v/v) USP-Grade Isopropyl Alcohol
Container Type Trigger Spray Bottle
Filtered Filtered
For Use With (Application) Removal of Process Residues, Routine Glove Decontamination
Quantity 12/Cs.
Volume (English) 22 oz.
Volume (Metric) 651 mL