missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Sephadex™ G-50, Medium
Description
Sephadex™ G-50 has been used to investigate effects from snake venom, isolate lectins from various species and to demonstrate the positive influence of 20% ethanol on proteinase desorption.
Specifications
Specifications
| Packaging | Poly Bottle |
| Quantity | 50 g |
| Product Line | Sephadex |
| MDL Number | MFCD00132771 |
| CAS | 9004-54-0 |
Sephadex is a registered trademark of GE Healthcare