missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Sephadex™ G-25, Fine

Catalog No. 111015859
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015859 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015859 Supplier MilliporeSigma™ Supelco™ Supplier No. G258010G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sephadex™ G-25 is a gel filtration medium used in affinity chromatography, protein chromatography and gel filtration chromatography.

Sephadex™ is a beaded gel filtration medium prepared by cross-linking dextran with epichlorohydrin under alkaline conditions.

Specifications

Packaging Poly Bottle
Quantity 10 g
Product Line Sephadex
MDL Number MFCD00132770
CAS 9041-35-4

Sephadex is a registered trademark of GE Healthcare