missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Sephadex™ G-25, Coarse

Catalog No. 111015860
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015860 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015860 Supplier MilliporeSigma™ Supelco™ Supplier No. G2530050G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sephadex™ G-25 is a gel filtration medium used in affinity chromatography, protein chromatography and gel filtration chromatography.

Sephadex™ G-25 has been used to study the acceleration of diabetic wound healing and the treatment of epithelial damage.

Specifications

Particle Size 0.10 to 0.3 mm
Technique Chromatography
Packaging Poly Bottle
Quantity 50 g
Product Line Sephadex
Material Cellulose
MDL Number MFCD00132770
CAS 9041-35-4

Sephadex is a registered trademark of GE Healthcare