missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Sephadex™ G-100, Medium

Catalog No. 111015867
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015867 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015867 Supplier MilliporeSigma™ Supelco™ Supplier No. G10012010G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sephadex™ G-100 is a gel filtration medium used in protein chromatography, affinity chromatography and gel filtration chromatography.

Sephadex™ is suitable for large-scale purification. It is considered stable and can be utilized more than once. Sephadex™ is composed of repeating glucose units attached by α-1,6 glucosidic bonds. This structure ensures more aptamer binding sites and greater binding capacity.

Specifications

Packaging Poly Bottle
Quantity 10 g
Product Line Sephadex
MDL Number MFCD00132235
CAS 9050-94-6

Sephadex is a registered trademark of GE Healthcare