Learn More
United Scientific™ Separatory Funnel
Separatory Funnels provide excellent clarity and strength.
Supplier: United Scientific™ 81203
Description
- These pear-shaped polypropylene separating funnels feature excellent clarity and strength, and include a leak-proof threaded screw cap
- The polypropylene stopcock is fitted with a self-lubricating PTFE plug
- Not graduated
Chemical Identifiers
| Polypropylene | |
| Clear | |
| Pear-shaped Separatory Funnel |
Specifications
| 90 mm | |
| Polypropylene | |
| 45 mm |
| 500 mL | |
| Clear | |
| Pear-shaped Separatory Funnel |