Learn More
EKF Micro12™ Centrifuge Tube Adapters
For use with Micro12 centrifuge
$198.96
Specifications
Type Centrifuge Tube Adapter
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
23550119
View Documents Promotion Details
EKF
230119
0.2mL thermal cycling tubes
Pack of 6 Each for $198.96
Only null left
23550121
View Documents Promotion Details
EKF
230121
0.5 and 0.6mL tubes
Pack of 6 Each for $198.96
Only null left
Description
Description
Specifications
Specifications
Centrifuge Tube Adapter