Learn More
Waters Limited Sep-Pak Aminopropyl (NH2) 6 cc Vac Cartridge, 1 g Sorbent per Cartridge, 55 - 105 m, 30/pk
Catalog No. 50785762
Encompass
Sep-Pak Amino Propyl (NH2) Vac cartridges contain a moderately polar, silica-based bonded phase with weakly basic surface. The sorbent can be used as a polar sorbent with different selectivity for acidic/basic analytes or as a weak anion exchanges in aqueous medium below pH 8. Applications include extraction of phenols and phenolic pigments, petroleum fractionation, saccharides and drugs and metabolites. These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.
Catalog No. 50785762 Supplier Waters Limited Supplier No. WAT054605
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Sep-Pak Amino Propyl (NH2) Vac cartridges contain a moderately polar, silica-based bonded phase with weakly basic surface. The sorbent can be used as a polar sorbent with different selectivity for acidic/basic analytes or as a weak anion exchanges in aqueous medium below pH 8. Applications include extraction of phenols and phenolic pigments, petroleum fractionation, saccharides and drugs and metabolites. These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.