missing translation for 'onlineSavingsMsg'
Learn More

ILC Dover Sentinel XT Breathing Tube

Catalog No. 19631109
Change view
Click to view available options
Size:
Regular
X-Long
Length (English):
25 in.
33 in.
Length (Metric):
63.5 cm
83.8 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (English) Length (Metric)
19631109 X-Long 25 in. 63.5 cm
19631108 Regular 33 in. 83.8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19631109 Supplier ILC Dover Supplier No. S31105
Only null left

Choose from two different sizes. For use with the Sentinel XT blower and clear hoods.

  • For use with Sentinel XT Blower and hoods
  • Two different sizes -- 25 in. or 33 in.

Specifications

Product Type Breathing Tube
Size X-Long
For Use With (Equipment) Sentinel XT Blower and Clear Hoods
Model S-3110-5
Quantity 5/Pk.
Length (English) 25 in.
Length (Metric) 63.5 cm