Learn More
ILC Dover Sentinel XL HP Half Hood System - Alkaline Battery Pack with Duffel
Ideal for healthcare settings, the Sentinel XL HP Clear Hood System includes S-4001 Clear Half Hood, 33 in. breathing Tube, Sentinel XL Blower, Flow Meter, Plug, alkaline battery pack, Quick Loc Belt, 2 HEPA filters and carrying tote.
Supplier: ILC Dover S300017
Description
- Ideal for use in Healthcare settings
- Sentinel XL powered air-purifying respirator assemblies and systems include parts required to build a full system or to replace existing parts
- Belt-mounted blower distributes ambient air through a filter to provide clean, breathable air to the wearer
- PAPRs allow workers to move freely and unrestricted
- Sentinel XL system parts are compatible with components of the same brand and series
Specifications
| PAPR System with Hood | |
| S-4001 Clear Half Hood, 33 in. Beathing Tube, Sentinel XL Blower, Flow Meter, Plug, alkaline battery pack, Quick Loc Belt, 2 HEPA filters and carrying tote | |
| Sentinel XL |