Learn More
ILC Dover Sentinel XL HP BioShield System - Rechargeable NIMH Battery with Duffel
Catalog No. 19631074
Change view
Click to view available options
Includes: Duffel Bag
Catalog No.
19631074 Includes: Duffel Bag
1 options
Catalog No. 19631074 Supplier ILC Dover Supplier No. S300022
Only null left
Ideal for healthcare settings, the Sentinel XL HP Clear Hood System includes S-2028 Full Hood, 33 in. breathing Tube, Sentinel XL Blower, Flow Meter, Plug, NiMH Rechargeable battery, charging cord, Quick Loc Belt, 2 HEPA filters and carrying tote.
  • Ideal for use in Healthcare settings
  • Sentinel XL powered air-purifying respirator assemblies and systems include parts required to build a full system or to replace existing parts
  • Belt-mounted blower distributes ambient air through a filter to provide clean, breathable air to the wearer
  • PAPRs allow workers to move freely and unrestricted
  • Sentinel XL system parts are compatible with components of the same brand and series
Specifications
Product Type PAPR System with Hood
Includes S-2028 Full Hood, 33 in. Beathing Tube, Sentinel XL Blower, Flow Meter, Plug, NiMH Rechargeable battery, charging cord, Quick Loc Belt, 2 HEPA filters and carrying tote
Model Sentinel XL