Learn More
ILC Dover Sentinel XL CBRN System - Rechargeable NIMH Battery with Duffel
Description
- Comes with Butyl Hood, Sentinel XL Blower, NiMH battery with charging cord, 3 CBRN cartridge filters and a duffel bag
- Sentinel XL powered air-purifying respirator assemblies and systems include parts required to build a full system or to replace existing parts
- Belt-mounted blower distributes ambient air through a filter to provide clean, breathable air to the wearer
- PAPRs allow workers to move freely and unrestricted
- Sentinel XL system parts are compatible with components of the same brand and series
Specifications
Specifications
| Product Type | PAPR System with Hood |
| Includes | Comes with Butyl Hood, Sentinel XL Blower, NiMH battery with charging cord, 3 CBRN cartridge filters and a duffel bag |
| Model | S-5000-01 |