Learn More
ILC Dover Sentinel XL CBRN System - Alkaline Battery Pack with Backpack
Catalog No. 19631091
Change view
Click to view available options
Includes: Backpack
Catalog No.
19631091 Includes: Backpack
1 options
Catalog No. 19631091 Supplier ILC Dover Supplier No. S500008
Only null left
The Sentinel XL CBRN PAPR system features an adjustable headband, is comfortable to wear, requires no fit-testing, and the lay flat design protects the system and allows it to stow easily in a backpack.
  • Comes with Butyl Hood, Sentinel XL Blower, alkaline battery back, flow meter, 3 CBRN cartridge filters and a backpack
  • Sentinel XL powered air-purifying respirator assemblies and systems include parts required to build a full system or to replace existing parts
  • Belt-mounted blower distributes ambient air through a filter to provide clean, breathable air to the wearer
  • PAPRs allow workers to move freely and unrestricted
  • Sentinel XL system parts are compatible with components of the same brand and series
Specifications
Product Type PAPR System with Hood
Includes Comes with Butyl Hood, Sentinel XL Blower, alkaline battery back, flow meter, 3 CBRN cartridge filters and a backpack
Model S-5000-08