missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One SensoPlate™ 96 Well Flat Bottomed Microplates with Lid
GREENER_CHOICE

Catalog No. 07001003
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07001003 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07001003 Supplier Greiner Bio-One Supplier No. 655892P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Case of 1

Allows transmission measurements in 350nm range.

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Optically clear borosilicate glass bottom with light path of 175±15μm, black polystyrene frame

Applications:

  • Fluorescence cor

Specifications

Format Standard
Lid With Lid
Array 8 x 12
Color Black
Material Polystyrene
No. of Wells 96
Sterility Sterile
Well Shape Flat Bottom (F-well)
Quantity Case of 1
Well Design Chimney