Learn More
Sensidyne™ RespiFit™ Respirator Qualitative Fit Test Kits
Catalog No. 19819028
Encompass
Change view
Click to view available options
Model:
7013304-1
Catalog No. Model
19819028 7013304-1
1 options
Catalog No. 19819028 Supplier Sensidyne™ Supplier No. 70133041
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Only Sensidyne 5100 smoke tubes and the RespiFit™ smoke tube pump (200 ml/min) are to be used with this fit test kit.
  • RespiFit Smoke Tube Pump
  • AA batteries
  • Box of smoke tubes
  • 30 in. tubing
  • 1 in. tube pieces
  • Smoke tube caps
  • Instructions
Specifications
Product Type Automatic Respiratory Qualitative Fit Test Kit
Includes 10 of No. 5100 smoke tubes, RespiFit smoke tube, pump, 30 in. of tubing, tube caps, Delivers 200mL/min., No hoods or test enclosures required
Model 7013304-1