Learn More
Sensidyne™ Airflow Indicator Kit
Catalog No. 19819031
Encompass
Change view
Click to view available options
Model:
7014094
Catalog No. Model
19819031 7014094
1 options
Catalog No. 19819031 Supplier Sensidyne™ Supplier No. 7014094
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Box of 10 Smoke Tubes, Aspirator Bulb, Tube Tip Breaker, Carrying Pouch
Offers a complete portable package for conducting qualitative respirator fit testing.
  • Highly recommended to track air flow currents
  • For fit testing respirators per OSHA 29 CFR part 1910.134, irritant fume protocol
Specifications
Product Type Airflow Indicator Kit
For Use With (Application) To track air flow currents and for fit testing respirators per OSHA 29 CFR part 1910.134, irritant fume protocol
Model 7014094